6-Bromo-2-((3-((tert-butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)methyl)furo[3,2-b]pyridine

Supplier: Matrix Scientific

054172-100MG 054172-500MG
101906-446EA 364.62 USD
101906-446 101906-448
6-Bromo-2-((3-((tert-butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)methyl)furo[3,2-b]pyridine
6-Bromo-2-((3-((tert-butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)methyl)furo[3,2-b]pyridine

Hazard: Irritant

Formula: C₁₉H₂₉BrN₂O₂Si
MDL Number: MFCD13563101
CAS Number: 1203499-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR