tert-Butyl (5-methyl-1H-pyrrolo[2,3-b]pyridin-3-yl)methylcarbamate

Supplier: Matrix Scientific

054168-100MG 054168-500MG
101906-430EA 364.62 USD
101906-430 101906-432
tert-Butyl (5-methyl-1H-pyrrolo[2,3-b]pyridin-3-yl)methylcarbamate
tert-Butyl (5-methyl-1H-pyrrolo[2,3-b]pyridin-3-yl)methylcarbamate

Hazard: Irritant

Formula: C₁₄H₁₉N₃O₂
MDL Number: MFCD13563097
CAS Number: 1198107-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR