Dimethyl-5-methyl-1H-pyrrolo[2,3-b]pyridine-1,3-dicarboxylate

Supplier: Matrix Scientific

054167-100MG 054167-500MG
101906-426EA 364.62 USD
101906-426 101906-428
Dimethyl-5-methyl-1H-pyrrolo[2,3-b]pyridine-1,3-dicarboxylate
Dimethyl-5-methyl-1H-pyrrolo[2,3-b]pyridine-1,3-dicarboxylate

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₄
MDL Number: MFCD13563096
CAS Number: 1198106-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR