(E)-5-Methyl-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde oxime

Supplier: Matrix Scientific

054163-100MG 054163-500MG
101906-410EA 364.62 USD
101906-410 101906-412
(E)-5-Methyl-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde oxime
(E)-5-Methyl-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde oxime

Hazard: Irritant

Formula: C₉H₉N₃O
MDL Number: MFCD13563092
CAS Number: 1198098-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR