(E)-Methyl-3-(7-chloro-1H-pyrrolo[2,3-c]pyridin-5-yl)acrylate

Supplier: Matrix Scientific

054162-100MG 054162-500MG
101906-406EA 364.62 USD
101906-406 101906-408
(E)-Methyl-3-(7-chloro-1H-pyrrolo[2,3-c]pyridin-5-yl)acrylate
(E)-Methyl-3-(7-chloro-1H-pyrrolo[2,3-c]pyridin-5-yl)acrylate

Hazard: Irritant

Formula: C₁₁H₉ClN₂O₂
MDL Number: MFCD13176654
CAS Number: 1198098-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR