4,5-Dichloro-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine

Supplier: Matrix Scientific

054160-100MG 054160-500MG
101906-398EA 364.62 USD
101906-398 101906-400
4,5-Dichloro-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine
4,5-Dichloro-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₁₆H₂₄Cl₂N₂Si
MDL Number: MFCD13176652
CAS Number: 685513-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR