1-(tert-Butoxycarbonyl)-5-methyl-1H-pyrrolo-[2,3-b]pyridine-3-carboxylic acid

Supplier: Matrix Scientific

054159-100MG 054159-500MG
101906-394EA 307.69 USD
101906-394 101906-396
1-(tert-Butoxycarbonyl)-5-methyl-1H-pyrrolo-[2,3-b]pyridine-3-carboxylic acid
1-(tert-Butoxycarbonyl)-5-methyl-1H-pyrrolo-[2,3-b]pyridine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₄
MDL Number: MFCD13176651
CAS Number: 1198097-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR