tert-Butyl-3-formyl-5-methyl-1H-pyrrolo[2,3-b]pyridine-1-carboxylate

Supplier: Matrix Scientific

054158-100MG 054158-500MG
101906-390EA 364.62 USD
101906-390 101906-392
tert-Butyl-3-formyl-5-methyl-1H-pyrrolo[2,3-b]pyridine-1-carboxylate
tert-Butyl-3-formyl-5-methyl-1H-pyrrolo[2,3-b]pyridine-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₃
MW: 260.3 g/mol
MDL Number: MFCD13176650
CAS Number: 1198097-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR