5-Methoxy-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine

Supplier: Matrix Scientific

054157-100MG 054157-500MG
101906-386EA 364.62 USD
101906-386 101906-388
5-Methoxy-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine
5-Methoxy-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₁₇H₂₈N₂OSi
MDL Number: MFCD13176649
CAS Number: 1198097-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR