Methyl-3-iodo-1-tosyl-1H-pyrrolo[2,3-b]pyridine-4-carboxylate
Supplier: Matrix Scientific
054154-100MG
054154-500MG
101906-374EA
364.62
USD
101906-374
101906-376
Methyl-3-iodo-1-tosyl-1H-pyrrolo[2,3-b]pyridine-4-carboxylate
Methyl-3-iodo-1-tosyl-1H-pyrrolo[2,3-b]pyridine-4-carboxylate
Hazard: Irritant
Formula:
C₁₆H₁₃IN₂O₄S |
MDL Number:
MFCD13176647 CAS Number: 1198097-28-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...