tert-Butyl-5-bromo-7-chloro-1H-pyrrolo[2,3-c]pyridine-1-carboxylate

Supplier: Matrix Scientific

054151-100MG 054151-500MG
101906-362EA 364.62 USD
101906-362 101906-364
tert-Butyl-5-bromo-7-chloro-1H-pyrrolo[2,3-c]pyridine-1-carboxylate
tert-Butyl-5-bromo-7-chloro-1H-pyrrolo[2,3-c]pyridine-1-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₂BrClN₂O₂
MW: 331.6 g/mol
MDL Number: MFCD13176644
CAS Number: 1198096-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR