tert-Butyl-5-bromo-7-chloro-3-iodo-1H-pyrrolo[2,3-c]pyridine-1-carboxylate

Supplier: Matrix Scientific

054150-100MG 054150-500MG
101906-358EA 364.62 USD
101906-358 101906-360
tert-Butyl-5-bromo-7-chloro-3-iodo-1H-pyrrolo[2,3-c]pyridine-1-carboxylate
tert-Butyl-5-bromo-7-chloro-3-iodo-1H-pyrrolo[2,3-c]pyridine-1-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₁BrClIN₂O₂
MW: 457.5 g/mol
MDL Number: MFCD13176643
CAS Number: 1198096-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR