5-Fluoro-1H-pyrrolo[2,3-b]pyridin-4-ol

Supplier: Matrix Scientific

054146-100MG 054146-500MG
101906-342EA 384.62 USD
101906-342 101906-344
5-Fluoro-1H-pyrrolo[2,3-b]pyridin-4-ol
5-Fluoro-1H-pyrrolo[2,3-b]pyridin-4-ol

Hazard: Irritant

Formula: C₇H₅FN₂O
MDL Number: MFCD13563089
CAS Number: 1203499-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR