5-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carboxylic acid

Supplier: Matrix Scientific

054144-100MG 054144-500MG
101906-334EA 315.38 USD
101906-334 101906-336
5-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carboxylic acid
5-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carboxylic acid

Hazard: Irritant

Formula: C₈H₅ClN₂O₂
MDL Number: MFCD13563088
CAS Number: 1203498-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR