5-Methyl-1H-pyrrolo[2,3-b]pyridine-3-carboxylic acid

Supplier: Matrix Scientific

054143-100MG 054143-500MG
101906-330EA 244.62 USD
101906-330 101906-332
5-Methyl-1H-pyrrolo[2,3-b]pyridine-3-carboxylic acid
5-Methyl-1H-pyrrolo[2,3-b]pyridine-3-carboxylic acid

Hazard: Irritant

Formula: C₉H₈N₂O₂
MDL Number: MFCD13563087
CAS Number: 1198095-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR