6-((Trimethylsilyl)ethynyl)-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one

Supplier: Matrix Scientific

054139-100MG 054139-500MG
101906-314EA 364.62 USD
101906-314 101906-316
6-((Trimethylsilyl)ethynyl)-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one
6-((Trimethylsilyl)ethynyl)-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₂Si
MW: 246.34 g/mol
MDL Number: MFCD13563083
CAS Number: 1203499-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR