6-(3-Hydroxyprop-1-ynyl)-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one

Supplier: Matrix Scientific

054138-100MG 054138-500MG
101906-310EA 364.62 USD
101906-310 101906-312
6-(3-Hydroxyprop-1-ynyl)-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one
6-(3-Hydroxyprop-1-ynyl)-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one

Hazard: Irritant

Formula: C₁₀H₈N₂O₃
MW: 204.19 g/mol
MDL Number: MFCD13563082
CAS Number: 1203499-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR