7-Iodo-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one

Supplier: Matrix Scientific

054136-100MG 054136-500MG
101906-302EA 364.62 USD
101906-302 101906-304
7-Iodo-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one
7-Iodo-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one

Hazard: Irritant

Formula: C₇H₅IN₂O₂
MDL Number: MFCD13176677
CAS Number: 1203499-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR