1-tert-Butyl-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one

Supplier: Matrix Scientific

054135-100MG 054135-500MG
101906-298EA 364.62 USD
101906-298 101906-300
1-tert-Butyl-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one
1-tert-Butyl-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₂
MDL Number: MFCD13176676
CAS Number: 1203499-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR