7-Bromo-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one
Supplier: Matrix Scientific
054132-100MG
054132-500MG
101906-290EA
135.38
USD
101906-290
101906-292
7-Bromo-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one
7-Bromo-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one
Hazard: Irritant
Formula:
C₇H₅BrN₂O₂ MW: 229.0308 g/mol |
MDL Number:
MFCD01648627 CAS Number: 105544-36-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...