8-Iodo-1-(4-methoxybenzyl)-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one

Supplier: Matrix Scientific

054127-100MG 054127-500MG
101906-270EA 364.62 USD
101906-270 101906-272
8-Iodo-1-(4-methoxybenzyl)-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one
8-Iodo-1-(4-methoxybenzyl)-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one

Hazard: Irritant

Formula: C₁₅H₁₃IN₂O₃
MW: 396.19 g/mol
MDL Number: MFCD13176669
CAS Number: 1203499-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR