1-Benzyl-7-iodo-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one

Supplier: Matrix Scientific

054126-100MG 054126-500MG
101906-266EA 364.62 USD
101906-266 101906-268
1-Benzyl-7-iodo-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one
1-Benzyl-7-iodo-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one

Hazard: Irritant

Formula: C₁₄H₁₁IN₂O₂
MDL Number: MFCD13176668
CAS Number: 1203499-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR