1-(4-Methoxybenzyl)-6-((trimethylsilyl)ethynyl)-1-(4-methoxybeH-pyrido[2,3-b][1,4]oxazin-2(3H)-one

Supplier: Matrix Scientific

054123-100MG 054123-500MG
101906-254EA 364.62 USD
101906-254 101906-256
1-(4-Methoxybenzyl)-6-((trimethylsilyl)ethynyl)-1-(4-methoxybeH-pyrido[2,3-b][1,4]oxazin-2(3H)-one
1-(4-Methoxybenzyl)-6-((trimethylsilyl)ethynyl)-1-(4-methoxybeH-pyrido[2,3-b][1,4]oxazin-2(3H)-one

Hazard: Irritant

Formula: C₂₀H₂₂N₂O₃Si
MW: 366.5 g/mol
MDL Number: MFCD13176665
CAS Number: 1198425-56-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR