tert-Butyl-7-iodo-2H-pyrido[3,2-b][1,4]oxazine-4(3H)-carboxylate

Supplier: Matrix Scientific

054122-100MG 054122-500MG
101906-250EA 278.46 USD
101906-250 101906-252
tert-Butyl-7-iodo-2H-pyrido[3,2-b][1,4]oxazine-4(3H)-carboxylate
tert-Butyl-7-iodo-2H-pyrido[3,2-b][1,4]oxazine-4(3H)-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₅IN₂O₃
MW: 362.17 g/mol
MDL Number: MFCD13176664
CAS Number: 1198108-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR