1H-Pyrido[2,3-b][1,4]oxazin-2(3H)-one
Supplier: Matrix Scientific
054120-100MG
054120-500MG
101906-246EA
156.92
USD
101906-246
101906-248
1H-Pyrido[2,3-b][1,4]oxazin-2(3H)-one
1H-Pyrido[2,3-b][1,4]oxazin-2(3H)-one
Hazard: Irritant
Formula:
C₇H₆N₂O₂ |
MDL Number:
MFCD01549954 CAS Number: 136742-83-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...