2-Amino-6-methyl-1,8-naphthyridine-3-carbonitrile

Supplier: Matrix Scientific

054119-100MG 054119-500MG
101906-242EA 364.62 USD
101906-242 101906-244
2-Amino-6-methyl-1,8-naphthyridine-3-carbonitrile
2-Amino-6-methyl-1,8-naphthyridine-3-carbonitrile

Hazard: Irritant

Formula: C₁₀H₈N₄
MDL Number: MFCD13563080
CAS Number: 1203499-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR