2-(Dimethoxymethyl)-6-methyl-1,8-naphthyridine
Supplier: Matrix Scientific
054118-100MG
054118-500MG
101906-238EA
364.62
USD
101906-238
101906-240
2-(Dimethoxymethyl)-6-methyl-1,8-naphthyridine
2-(Dimethoxymethyl)-6-methyl-1,8-naphthyridine
Hazard: Irritant
Formula:
C₁₂H₁₄N₂O₂ |
MDL Number:
MFCD13563079 CAS Number: 1203499-56-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...