(E)-Methyl-3-(2-fluoro-6-(pyrrolidin-1-yl)pyridin-3-yl)acrylate

Supplier: Matrix Scientific

054115-100MG 054115-500MG
101906-226EA 364.62 USD
101906-226 101906-228
(E)-Methyl-3-(2-fluoro-6-(pyrrolidin-1-yl)pyridin-3-yl)acrylate
(E)-Methyl-3-(2-fluoro-6-(pyrrolidin-1-yl)pyridin-3-yl)acrylate

Hazard: Irritant

Formula: C₁₃H₁₅FN₂O₂
MDL Number: MFCD13563076
CAS Number: 1203500-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR