1-(5-Bromo-2-chloropyridin-3-yl)ethanone

Supplier: Matrix Scientific

054114-100MG 054114-500MG
101906-222EA 43.08 USD
101906-222 101906-224
1-(5-Bromo-2-chloropyridin-3-yl)ethanone
1-(5-Bromo-2-chloropyridin-3-yl)ethanone

Hazard: Irritant

Formula: C₇H₅BrClNO
MDL Number: MFCD08436181
CAS Number: 886365-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR