3-Allyl-2-fluoro-6-(pyrrolidin-1-yl)pyridine

Supplier: Matrix Scientific

054112-100MG 054112-500MG
101906-214EA 364.62 USD
101906-214 101906-216
3-Allyl-2-fluoro-6-(pyrrolidin-1-yl)pyridine
3-Allyl-2-fluoro-6-(pyrrolidin-1-yl)pyridine

Hazard: Irritant

Formula: C₁₂H₁₅FN₂
MDL Number: MFCD13563074
CAS Number: 1203499-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR