2-Fluoro-6-(pyrrolidin-1-yl)nicotinic acid

Supplier: Matrix Scientific

054111-100MG 054111-500MG
101906-210EA 364.62 USD
101906-210 101906-212
2-Fluoro-6-(pyrrolidin-1-yl)nicotinic acid
2-Fluoro-6-(pyrrolidin-1-yl)nicotinic acid

Hazard: Irritant

Formula: C₁₀H₁₁FN₂O₂
MDL Number: MFCD13563073
CAS Number: 1203499-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR