2-Fluoro-6-(pyrrolidin-1-yl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Supplier: Matrix Scientific
054110-100MG
054110-500MG
101906-206EA
364.62
USD
101906-206
101906-208
2-Fluoro-6-(pyrrolidin-1-yl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
2-Fluoro-6-(pyrrolidin-1-yl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Hazard: Irritant
Formula:
C₁₅H₂₂BFN₂O₂ |
MDL Number:
MFCD13563072 CAS Number: 1309978-63-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...