2-Fluoro-6-(pyrrolidin-1-yl)nicotinaldehyde

Supplier: Matrix Scientific

054109-100MG 054109-500MG
101906-202EA 315.38 USD
101906-202 101906-204
2-Fluoro-6-(pyrrolidin-1-yl)nicotinaldehyde
2-Fluoro-6-(pyrrolidin-1-yl)nicotinaldehyde

Hazard: Irritant

Formula: C₁₀H₁₁FN₂O
MDL Number: MFCD13563071
CAS Number: 1203499-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR