2-Methoxy-5-methylnicotinonitrile

Supplier: Matrix Scientific

054106-100MG 054106-500MG
101906-190EA 364.62 USD
101906-190 101906-192
2-Methoxy-5-methylnicotinonitrile
2-Methoxy-5-methylnicotinonitrile

Hazard: Irritant

Formula: C₈H₈N₂O
MDL Number: MFCD13563068
CAS Number: 149379-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR