2-(3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-5-fluoro-3-iodopyridine

Supplier: Matrix Scientific

054104-100MG 054104-500MG
101906-182EA 364.62 USD
101906-182 101906-184
2-(3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-5-fluoro-3-iodopyridine
2-(3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-5-fluoro-3-iodopyridine

Hazard: Irritant

Formula: C₁₆H₂₆FIN₂OSi
MDL Number: MFCD13563066
CAS Number: 1203499-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR