tert-Butyl (2-chloro-5-methylpyridin-3-yl)methylcarbamate

Supplier: Matrix Scientific

054102-100MG 054102-500MG
101906-174EA 364.62 USD
101906-174 101906-176
tert-Butyl (2-chloro-5-methylpyridin-3-yl)methylcarbamate
tert-Butyl (2-chloro-5-methylpyridin-3-yl)methylcarbamate

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O₂
MDL Number: MFCD13563064
CAS Number: 1203499-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR