1-(2-Chloro-5-fluoropyridin-3-yl)ethanone

Supplier: Matrix Scientific

054100-100MG 054100-500MG
101906-166EA 364.62 USD
101906-166 101906-168
1-(2-Chloro-5-fluoropyridin-3-yl)ethanone
1-(2-Chloro-5-fluoropyridin-3-yl)ethanone

Hazard: Irritant

Formula: C₇H₅ClFNO
MDL Number: MFCD13563063
CAS Number: 1203499-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR