3-(2-Aminopyridin-3-yl)propan-1-ol

Supplier: Matrix Scientific

054099-100MG 054099-500MG
101906-162EA 476.92 USD
101906-162 101906-164
3-(2-Aminopyridin-3-yl)propan-1-ol
3-(2-Aminopyridin-3-yl)propan-1-ol

Hazard: Irritant

Formula: C₈H₁₂N₂O
MDL Number: MFCD09927103
CAS Number: 89226-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR