2-Chloro-3-(dimethoxymethyl)-5-methylpyridine

Supplier: Matrix Scientific

054098-100MG 054098-500MG
101906-158EA 364.62 USD
101906-158 101906-160
2-Chloro-3-(dimethoxymethyl)-5-methylpyridine
2-Chloro-3-(dimethoxymethyl)-5-methylpyridine

Hazard: Irritant

Formula: C₉H₁₂ClNO₂
MDL Number: MFCD13563062
CAS Number: 1203499-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR