3-Amino-5-bromopyridin-4-ol

Supplier: Matrix Scientific

Synonyms: 3-Amino-5-bromo-4-hydroxypyridine

054096-100MG 054096-500MG
101906-150EA 69.23 USD
101906-150 101906-152
3-Amino-5-bromopyridin-4-ol
3-Amino-5-bromopyridin-4-ol

Hazard: Irritant

Formula: C₅H₅BrN₂O
MW: 189.012 g/mol
MDL Number: MFCD08692340
CAS Number: 101084-20-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR