2-Methoxy-5-methylnicotinaldehyde

Supplier: Matrix Scientific

Synonyms: 2-Methoxy-5-methylpyridine-3-carboxaldehyde

054095-100MG 054095-500MG
101906-146EA 364.62 USD
101906-146 101906-148
2-Methoxy-5-methylnicotinaldehyde
2-Methoxy-5-methylnicotinaldehyde

Hazard: Irritant

Formula: C₈H₉NO₂
MW: 151.17 g/mol
MDL Number: MFCD13563060
CAS Number: 1203499-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR