2-Chloro-5-methyl-3-(trimethylsilyl)pyridine
Supplier: Matrix Scientific
Synonyms:
2-Chloro-5-methyl-3-(trimethylsilyl)pyridine
054093-100MG
054093-500MG
101906-138EA
364.62
USD
101906-138
101906-140
2-Chloro-5-methyl-3-(trimethylsilyl)pyridine
2-Chloro-5-methyl-3-(trimethylsilyl)pyridine
Hazard: Irritant
Formula:
C₉H₁₄ClNSi MW: 199.76 g/mol |
MDL Number:
MFCD13563058 CAS Number: 1203498-92-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...