Methyl-2-chloro-5-methylnicotinate

Supplier: Matrix Scientific

Synonyms: Methyl 2-chloro-5-methylpyridine-3-carboxylate, Methyl 2-chloro-5-methylnicotinate

054092-100MG 054092-500MG
101906-134EA 75.38 USD
101906-134 101906-136
Methyl-2-chloro-5-methylnicotinate
Methyl-2-chloro-5-methylnicotinate

Hazard: Irritant

Formula: C₈H₈ClNO₂
MDL Number: MFCD07375372
CAS Number: 65169-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR