(E)-2-Chloro-5-methylnicotinaldehyde oxime

Supplier: Matrix Scientific

054091-100MG 054091-500MG
101906-130EA 375.38 USD
101906-130 101906-132
(E)-2-Chloro-5-methylnicotinaldehyde oxime
(E)-2-Chloro-5-methylnicotinaldehyde oxime

Hazard: Irritant

Formula: C₇H₇ClN₂O
MDL Number: MFCD13563057
CAS Number: 1203500-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR