2-Chloro-5-methylnicotinonitrile

Supplier: Matrix Scientific

054090-100MG 054090-500MG
101906-126EA 315.38 USD
101906-126 101906-128
2-Chloro-5-methylnicotinonitrile
2-Chloro-5-methylnicotinonitrile

Hazard: Irritant

Formula: C₇H₅ClN₂
MDL Number: MFCD07375374
CAS Number: 66909-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR