(E)-Methyl-3-(2-chloro-5-methylpyridin-3-yl)acrylate

Supplier: Matrix Scientific

054089-100MG 054089-500MG
101906-122EA 315.38 USD
101906-122 101906-124
(E)-Methyl-3-(2-chloro-5-methylpyridin-3-yl)acrylate
(E)-Methyl-3-(2-chloro-5-methylpyridin-3-yl)acrylate

Hazard: Irritant

Formula: C₁₀H₁₀ClNO₂
MDL Number: MFCD13563056
CAS Number: 1198401-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR