(E)-Methyl-3-(2-aminopyridin-3-yl)acrylate

Supplier: Matrix Scientific

054087-100MG 054087-500MG
101906-114EA 364.62 USD
101906-114 101906-116
(E)-Methyl-3-(2-aminopyridin-3-yl)acrylate
(E)-Methyl-3-(2-aminopyridin-3-yl)acrylate

Hazard: Irritant

Formula: C₉H₁₀N₂O₂
MDL Number: MFCD13563054
CAS Number: 1203500-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR