tert-Butyl-3-((3-iodo-5-methylpyridin-2-yloxy)methyl)pyrrolidine-1-carboxylate

Supplier: Matrix Scientific

054085-100MG 054085-500MG
101906-106EA 364.62 USD
101906-106 101906-108
tert-Butyl-3-((3-iodo-5-methylpyridin-2-yloxy)methyl)pyrrolidine-1-carboxylate
tert-Butyl-3-((3-iodo-5-methylpyridin-2-yloxy)methyl)pyrrolidine-1-carboxylate

Hazard: Irritant

Formula: C₁₆H₂₃IN₂O₃
MW: 418.28 g/mol
MDL Number: MFCD13563052
CAS Number: 1203499-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR