2-Chloro-5-methyl-3-[(trimethylsilyl)ethynyl]pyridine

Supplier: Matrix Scientific

054083-100MG 054083-500MG
101906-098EA 364.62 USD
101906-098 101906-100
2-Chloro-5-methyl-3-[(trimethylsilyl)ethynyl]pyridine
2-Chloro-5-methyl-3-[(trimethylsilyl)ethynyl]pyridine

Hazard: Irritant

Formula: C₁₁H₁₄ClNSi
MW: 223.78 g/mol
MDL Number: MFCD13563050
CAS Number: 1203499-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR