1-(5,6-Dimethoxypyridin-2-yl)ethanone

Supplier: Matrix Scientific

054082-100MG 054082-500MG
101906-094EA 253.85 USD
101906-094 101906-096
1-(5,6-Dimethoxypyridin-2-yl)ethanone
1-(5,6-Dimethoxypyridin-2-yl)ethanone

Hazard: Irritant

Formula: C₉H₁₁NO₃
MDL Number: MFCD13563049
CAS Number: 1203499-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR